Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

Z515825

Sigma-Aldrich

Schlenk Kjeldahl reaction and storage flask

capacity 250 mL, male joint: ST/NS 14/20

Synonym(s):

schlenk flasks, schlenk tubes, storage tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

material

borosilicate glass
pear-shaped
single-neck

Joint

male joint: ST/NS14/20

sterility

non-sterile

feature

closure type Female Joint: ST/NS & screw cap

packaging

pack of 1

manufacturer/tradename

Sigma-Aldrich

capacity

250 mL

Looking for similar products? Visit Product Comparison Guide

General description

2 mm glass stopcock. The side-arm accommodates a septum (use Z565695 or Z565679). Designed for small-scale manipulation of air and moisture-sensitive reagents. Thermoplastic screw caps with Viton O-rings usable to 140 deg C.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service