Key Documents
Z509353
Sigma-Aldrich
Weighing funnel
size L, anti-static PP
Synonym(s):
Weighing Apparatus, Weighing Device
Select a Size
1 PKG
$128.00
$128.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
$128.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.71
$128.00
Please contact Customer Service for Availability
material
anti-static PP
size
L
packaging
pkg of 20 ea
General description
  • Sit flat on balance - No rolling, no spilling!
  • Pouring necks designed to fit volumetric flasks, micro-tubes or any narrow neck apparatus for spill free transfer
  • Residual sample can be rinsed to guarantee 100% transfer
  • Suitable for safe handling of hazardous materials
  • Available in five different sizes: extra small, small, medium, large, and extra large
Choose from one of the most recent versions:
Certificates of Analysis (COA)