Documents
Z506699
Sigma-Aldrich
Aldrich® macro-micro KBr pellet die
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
technique(s)
IR spectroscopy: suitable
diam.
13 mm
General description
  • For producing 13mm diameter KBr pellets. Use in conjunction with a 12 ton lab press and a vacuum pump, capable of reducing pressure to 2mm, to reduce moisture in the sample. Heavy paper inserts in a variety of smaller apertures are available separately for making micropellets with the 13mm die set and also provide a useful frame for mounting and storing the pellets.
  • Hardened stainless steel piston and lower section
  • Black anodized aluminum body
  • Optically polished anvils
  • Viton® seals
  • Evacuable via 1/4in. O.D. hose connection
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Visit the Document Library
Customers Also Viewed
Slide 1 of 1