Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z420409

Medium porous filter plate for Table-Top Buchner funnels

for Z420379

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,380.00

$1,380.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$1,380.00

About This Item

UNSPSC Code:
41104925
NACRES:
NB.43

$1,380.00


Please contact Customer Service for Availability

material

high-density polyethylene

sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art H91462-4345

filter diam.

18 in.

color

none

pore size

45—90 μm

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service