Key Documents
Z418560
Sigma-Aldrich
System 45 vessel with threaded side port and removable fritted disc
capacity ~20 mL, Complete
About This Item
UNSPSC Code:
12352200
NACRES:
NB.43
L × diam.
50 mm × 28 mm
capacity
~20 mL
General description
Cylindrical style vessel has a 2 mm PTFE valve at the top, with a GL 18 threaded side port. The 32 mm threaded bottom valve assembly incorporates System 45 technology. The coarse fritted disc is easily removed from the PTFE adapter for cleaning and/or replacement. PTFE top and bottom valves require 1/4-turn to open or close. Valve stems have a 1/4-28 internal thread in the stem which allows a variety of adapters to be used. For male 1/4-28 and male Luer adapters, see Cat. No. Z417416 and Z417424.
Legal Information
System 45 is a trademark of John Babashak
Choose from one of the most recent versions:
Certificates of Analysis (COA)