Key Documents
Z414123
Sigma-Aldrich
Aldrich® offset enlarging adapter
male joint: ST/NS 24/40, female joint: ST/NS 34/45
Synonym(s):
glass adapter, glass adapters
Select a Size
1 EA
$144.00
$144.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$144.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$144.00
Please contact Customer Service for Availability
Joint
female joint: ST/NS34/45
male joint: ST/NS24/40
Related Categories
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)