Documents
Z413569
Sigma-Aldrich
Aldrich® three-way adapter with thermometer joint
joint: ST/NS 19/22
Synonym(s):
adapter laboratory, glass adapter, glass adapters
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
thermometer joint ST/NS10/18
joint: ST/NS19/22
Related Categories
General description
A 50mm immersion length thermometer is recommended.
Side-arm is at 75° angle from bottom joint. Distance from joint to side-arm is 75mm.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC