Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z413100

Sigma-Aldrich

Aldrich® micro distilling splitter head

Unjacketed, Overall H 260 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104812
NACRES:
NB.43
Pricing and availability is not currently available.

Overall H

260 mm

immersion depth

50 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The column and receiver joints are 14/20 with a 10/18 thermometer joint, two 2 mm glass stopcocks, PTFE 0-3 mm metering valve. Removable condenser cold-finger for easy cleaning.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. can you tell what cone dimension is this equipment ? Z413100 - micro distilling splitter head : 14/23 , 24/29 , 29/32 ?

    1 answer
    1. The column and receiver joints are 14/20 with a 10/18 thermometer joint, two 2 mm glass stopcocks, PTFE 0-3 mm metering valve. Removable condenser cold-finger for easy cleaning. The overall height is 260 mm and the immersion depth is 50 mm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service