Key Documents
Z409324
Sigma-Aldrich
Aldrich® Schlaker reaction tube
capacity 100 mL, screw-thread joint
Select a Size
1 EA
$537.00
$537.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$537.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$537.00
Please contact Customer Service for Availability
material
colorless tube
glass tube
round bottom
sterility
non-sterile
feature
graduations: no
neck thread PUSH FIT (14/20 top joint)
capacity
100 mL
binding type
non-treated surface
General description
The glass frit in the side-arm permits filtration of excess reagents and solvent removal with complete recovery of the resin for the next reaction. The "Schlaker" tube (derived from "Schlenk" and peptide "shaker") is designed for running oxygen and moisture sensitive reactions on a solid-phase resin.
Made of heavy-wall glass, with glass stopcock and frit (porosity 25-50 mm) on side-arm, and a plain or a threaded 14/20 top joint. Compatible with Schlenk glassware. Compatible with the Aldrich All-in-One Glassware Kit.
Schlaker Tube Technique
1. Place resin in the tube
2. Evacuate and purge with N2 to remove oxygen via a rubber septum or a septum-inlet adapter in the top joint
3. Stir gently while heating in an oil bath for high temperatures, or place on a shaker for room temperature reactions
4. When the reaction is complete, apply positive N2 pressure through the septum in the top joint, and vacuum to the side-arm
5. Tilt the Schlaker tube on its side to allow solvent and excess reagents to flow out, leaving the resin behind
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)