Key Documents
Z407674
Sigma-Aldrich
Quartz wall clock-thermometer-hygrometer
About This Item
UNSPSC Code:
41122411
General description
Useful for the laboratory, office, or computer room. Measures RH from 0 to 100% in 2% subdivisions and temperature in °F. Attractive 93/4 in. diameter black face with fluorescent numerals makes it an easy-to-read addition to your office. Requires one AA battery (not included).
Choose from one of the most recent versions:
Certificates of Analysis (COA)