Key Documents
Z405760
Cotton blend glove liners
size S/M (womens)
Synonym(s):
fabric glove insert
Select a Size
1 PKG
$32.70
$32.70
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
$32.70
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
$32.70
Please contact Customer Service for Availability
material
cotton
white gloves
size
S/M (womens)
sterility
non-sterile
feature
latex free
powder-free
reusable
packaging
pack of 12 pair
manufacturer/tradename
Sigma-Aldrich
L
9 in.
Related Categories
General description
Use these gloves for product protection during inspection or light manufacturing. Also ideal for use as a liner underneath outer gloves. Reversible pattern allows glove to be worn on either hand. Lightweight styles feature unhemmed cuffs.
Choose from one of the most recent versions:
Certificates of Analysis (COA)