Key Documents
Z308854
Quickfit® conical flasks
capacity 250 mL, joint: ST/NS 34/35
Synonym(s):
FE250/5
About This Item
UNSPSC Code:
41121804
material
borosilicate glass
conical flask
Joint
joint: ST/NS34/35
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 5
manufacturer/tradename
Quickfit FE250/5
availability
not available in USA and Canada
capacity
250 mL
General description
With ground-glass socket for general use. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited