Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z308579

Quickfit® Drechsel bottles

capacity 125 mL

Synonym(s):

MF29/3/125

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.51
Product Z308579 is not currently sold in your country. Contact Technical Service

Joint

joint 24/29

manufacturer/tradename

Quickfit MF 29/3/125

availability

not available in USA and Canada

H

155 mm

capacity

125 mL

Looking for similar products? Visit Product Comparison Guide

General description

Select bottle and corresponding head. Note different bottles sizes are different heights.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service