Key Documents
Z302406
Quickfit® Cone/Screw-thread Adapters
18 thread (couples 7.0 - 8.5 mm diameter), joint: ST/NS 24/29
Synonym(s):
ST53/18
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Product Z302406 is not currently sold in your country. Contact Technical Service
Joint
joint: ST/NS24/29
packaging
pack of 2 ea
manufacturer/tradename
Quickfit ST 53/18
availability
not available in USA and Canada
not available in USA and Canada
fitting
18 thread (couples 7.0 - 8.5 mm diameter)
General description
The Quickfit screw-thread joints on these cone adapters provide a versatile means of coupling thermometers, bleed tubes, electrodes, etc; with Quickfit glassware. The joints are readily adjustable for depth of immersion by slackening the screw cap, and give excellent results under vacuum.
Can be used in apparatus at temperatures up to 300°C providing excessive heat is not applied directly to the screw-thread union.
For replacement parts, see related products section of web site listing.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)