Key Documents
Z302198
Tyvek® Pro.Tech total body coveralls
size XXL
About This Item
UNSPSC Code:
41100000
size
XXL
manufacturer/tradename
Tyvek
availability
not available in USA and Canada
General description
With full length zipper, elasticated wrist holes, integral elasticated hood and overshoes for protection in controlled, clean environments. Provides CE type 5 and Type 6 (particle and limited splash) protection.
Legal Information
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)