Key Documents
Z288225
KNF Laboport® mini-pump
Pump with vacuum gauge and regulator, AC/DC input 115 V AC
Synonym(s):
laboratory vacuum pump, mini diaphragm vacuum pump, mini vacuum pump, vacuum pump
About This Item
UNSPSC Code:
41105101
NACRES:
NB.79
feature
CE compliant
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
KNF
availability
not available in EU
parameter
35 psig max. pressure
5.5 L/min pumping speed
~120 torr vacuum
weight
4.2 lb
General description
The ideal pump for vacuum and pressure filtration, solid phase extraction, and blotting. High performance diaphragm vacuum/pressure pumps for moderately corrosive applications and any filtration or procedure that requires clean evacuation, transfer and compression of air, gases, and vapors. Pumps are available with gauges and regulators for precise control of vacuum and pressure.
  • Portable
  • Oil-free
  • Quiet operation
  • Maintenance-free, Ryton® pump head
  • Molded PTFE diaphragm
  • Kalrez® multi-port valves
  • Single stage
Legal Information
Kalrez is a registered trademark of E. I. du Pont de Nemours and Company
Laboport is a registered trademark of KNF Neuberger GmbH
Ryton is a registered trademark of Chevron Phillips Chemical Company LP
Choose from one of the most recent versions:
Certificates of Analysis (COA)