Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z286389

Sigma-Aldrich

All-polyethylene acid storage cabinet

large capacity

Synonym(s):

chemical storage cabinet, chemical storage cabinets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

manufacturer/tradename

Justrite 24160

H × W × D

35 in. × 36 in. × 22 in.

weight

110 lb

Looking for similar products? Visit Product Comparison Guide

General description

  • Stores up to 83 L
  • Two adjustable PE shelves
  • Storage in both doors for small bottles and flasks
  • Fits under counter or free-standing
These unique, PE storage cabinets feature one-piece, molded construction for maximum corrosion resistance and protection against leaks. Recessed tops permit use as free-standing workstations. Inside spill trays remove for easy cleaning. Can be padlocked to prevent unauthorized access.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service