Key Documents
Z278688
Sigma-Aldrich
Flow control pinch valve
Set of three
About This Item
UNSPSC Code:
40141600
NACRES:
NB.79
material
Delrin® body
General description
For tubing O.D. sizes 5/32 - 1/4 in, 5/16 - 3/8 in and 7/16 - 1/2 in.
Metered scale on valve body adjusts fluid from zero to full flow without fluid contact with valve. Calibration is accurate and repeatable. Valve mounts easily on existing soft wall tube lines without fittings or cutting.
Legal Information
Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)