Key Documents
Z268518
Sigma-Aldrich
Vacushield
in-line filter designed to be placed in front of a vacuum pump, pkg of 3 ea
About This Item
UNSPSC Code:
41104900
packaging
pkg of 3 ea
manufacturer/tradename
Gelman 4402
General description
Disposable, hydrophobic, in-line filter designed to be placed in front of a vacuum pump. Protects the pump from damage caused by aqueous solutions. Hose barb connector. Autoclavable.
Legal Information
Vacushield is a trademark of Pall Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)