Key Documents
Z268429
Sigma-Aldrich
Pall reusable syringe filter holders
diam. 25 mm
About This Item
UNSPSC Code:
41104900
eCl@ss:
32110206
material
polyacetal (resin construction)
polypropylene housing
sterility
non-sterile
packaging
pkg of 6 ea
manufacturer/tradename
Gelman 4320
diam.
25 mm
pore size
(N/A)
fitting
Luer inlet and outlet
General description
Reusable plastic filter holders provide an affordable alternative to disposable syringe filters. Autoclavable.
Choose from one of the most recent versions:
Certificates of Analysis (COA)