Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z260991

Nalgene® laboratory notebooks

standard paper pages, 1/4 in. grid

Synonym(s):

experiment notebook, lab notebook

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
AB.01

publication info

, 96 pp., hard cover

feature

1/4 in. grid

packaging

pack of 6 ea

manufacturer/tradename

Nalgene 6301-1000

color

forest green Cover

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • The Nalgene® laboratory notebooks feature durable pages with either a grid or horizontal lines.
  • The pages also contain spaces for dates, signatures of workers, and witnesses for securing patent protection.
  • The book includes pages for instructions on keeping research records, a table of contents, and details of notebook issuing.
  • The polyethylene cover is waterproof.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service