Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z257761

Enviro-Safe® pocket thermometer

greener alternative

temperature -10-110 °C, Without armor

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$52.10

$52.10


Available to ship onApril 17, 2025Details



Select a Size

Change View
1 EA
$52.10

About This Item

UNSPSC Code:
41112303
NACRES:
NB.79

$52.10


Available to ship onApril 17, 2025Details


feature

subdivision 1 °C

packaging

pack of 1 ea

manufacturer/tradename

HB Instruments B60570-1300

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

temperature measuring range

-10-110 °C

L

145 mm

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Total immersion. With green, non-hazardous, liquid fill. 160 mm L with aluminum case. An armor option is available, providing extra shielding for the thermometer.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is designed as a safer chemical without mercury. Click here for more information.

Legal Information

Enviro-Safe is a registered trademark of H-B Instrument Co.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service