Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z256641

Kontes® Williamson ST 14/20 macroscale kit

polyethylene case with foam liner

Synonym(s):

269330-0000

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.43

manufacturer/tradename

Kimble®

General description

Components, Qty
A
Conical flask, 15mL, 1
B Claisen adapter, 1
C Connecting adapter, 1
D Vacuum adapter, 1
E Water-jacketed condenser, 1
F Distilling column, 1
G Distillation receiver, 5mL, 1
H Kjeldahl-shaped flask, 50mL, 1
I Kjeldahl-shaped flask, 100mL, 1
J Separatory funnel, 125mL, 1
K Centrifuge/sublimation tube, 15mL, 1
L Stopcock, one-way, PE/nylon, 1
M Chromatography column, 1
N Polyacetal clamp, size 14, 4
O Filter adapter, Pluro stopper, 1
P Filter flask, 25mL, 1
Q Hirsch funnel, 1
R Hex-head stopper, 14/20, 1

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences
Kontes is a registered trademark of Kimble Kontes Asset Mgmt. Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service