Skip to Content
MilliporeSigma
All Photos(2)

Documents

Z248029

Sigma-Aldrich

Syringe PP/PE without needle

luer lock tip, centered, capacity 10 mL, graduated, 0.5 mL, sterile

Synonym(s):

Luer lock syringes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600
eCl@ss:
32070500
NACRES:
NB.03

sterility

sterile

feature

graduated 0.5 mL

packaging

pkg of 100 ea individually wrapped

capacity

10 mL

Looking for similar products? Visit Product Comparison Guide

General description

These polypropylene syringes with Luer tips, may be used with any size needle. The rigid polyethylene plungers of these disposable syringes are slightly oversized, creating a tight seal against the walls of the semi-flexible PP barrel, as well as providing an exceptionally smooth draw. No contaminating lubricants added.

Syringes are individually packaged (sterile) except for bulk packed products; these are supplied in plastic bags (non-sterile).

Order needles separately.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Sadeesh et al.
Cytotechnology, 68(6), 2271-2285 (2016-09-22)
The study examined the effects of different environmental stress on developmental competence and the relative abundance (RA) of various gene transcripts in oocytes and embryos of buffalo. Oocytes collected during cold period (CP) and hot period (HP) were matured, fertilized
E M Sadeesh et al.
Cytotechnology, 69(2), 289-305 (2017-01-11)
This study examined the effects of buffalo oocyte extracts (BOE) on donor cells reprogramming and molecular characterisation of oocytes screened via brilliant cresyl blue (BCB) staining and comparison of gene expression profiles of developmentally important genes in blastocysts from IVF
Karolina Janeczek Portalska et al.
Journal of tissue engineering and regenerative medicine, 10(5), 363-373 (2013-04-18)
Modular tissue engineering is a strategy to create scalable, self-assembling, three-dimensional (3D) tissue constructs. This strategy was used to deliver endothelial-like cells derived from bone marrow mesenchymal stromal cells (EL-MSCs) to locally induce vascularization. First, tissue engineered modules were formed
Sayed-Hadi Mirmalek-Sani et al.
Biomaterials, 34(22), 5488-5495 (2013-04-16)
Emergent technologies of regenerative medicine have the potential to overcome the limitations of organ transplantation by supplying tissues and organs bioengineered in the laboratory. Pancreas bioengineering requires a scaffold that approximates the biochemical, spatial and vascular relationships of the native

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service