Key Documents
Z238074
Sigma-Aldrich
Monogoggles
green Frame
About This Item
UNSPSC Code:
46181802
availability
not available in EU
color
green Frame
General description
Exclusive two-piece frame design. A separate nose piece of soft, pliable plastic molds to fit a variety of facial contours, providing better contact and a tighter seal around the nose and cheeks. Lightweight, transparent frames have an indirect ventilation system built-in to create an airflow across the lens, minimizing condensation. Lenses are made from clear, 0.060 in. thick, optical grade PC and are coated with FogGard to reduce fogging. Padded felt frames improve fit for workers with smaller facial features. Meets ANSI Z87.1 and CSA Z94.3 requirements. SEI certified.
Choose from one of the most recent versions:
Certificates of Analysis (COA)