Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z232211

Duran® graduated laboratory bottles, without caps

capacity 1,000 mL, amber glass bottle

Synonym(s):

218065402

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17
To inquire about this product Z232211, please contact your local MilliporeSigma office or dealer. Contact Technical Service

material

amber glass bottle
round bottle

Joint

Joints threaded neck (GL45)

sterility

non-sterile

feature

graduations
without cap

packaging

pack of 1 ea

manufacturer/tradename

DURAN® 21 806 54

capacity

1,000 mL

diam. × H

101 mm × 225 mm

Looking for similar products? Visit Product Comparison Guide

General description

DURAN® Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, amber, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The addition of a fused, external amber layer ensures that light sensitive contents are protected from the effects of photodegradation. The protection against ultra-violet light meets international USP/EP standards and lasts the lifetime of the bottle. The amber bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ Original amber bottles are supplied with or without iconic blue polypropylene cap and drip-free pouring ring for use up to 140 °C. All components are fully autoclavable at 121 or 134 °C.
Reusable bottles without caps or pouring rings for the storage of reagents, chemicals, solutions, tissue culture media, etc. Made of Duran borosilicate glass, Type 3.3, for maximum chemical and thermal shock resistance.

Optional pouring rings prevent drips when pouring from these bottles.

Application

Duran™ graduated laboratory bottles, without caps are suitable for the storage of reagents, chemicals, solutions, tissue culture media, and more.
Duran® graduated laboratory bottles, without caps are suitable for the storage of reagents, chemicals, solutions, tissue culture media, and more.

Features and Benefits

  • The Duran® graduated laboratory bottles are made of Duran borosilicate glass, Type 3.3, for maximum chemical and thermal shock resistance.
  • These bottles are reusable and autoclavable at 121 or 134 °C.
  • The optional pouring rings prevent drips when pouring from these bottles.
  • The bottles provide an easy-to-read scale with graduation marks to estimate the content.
  • They are suitable for long-term storage.
  • Manufactured from ambered DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • USP <660> (Spectral Transmission) and EP (3.2.1) compliant. Less than 10% spectral transmission between 290 nm and 450 nm.
  • The only external application of the colour ensures that internal surfaces have standard DURAN® glass properties
  • Very durable and chemically resistant amber colour
  • Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic print
  • Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service