Key Documents
Z220388
Sigma-Aldrich
Aldrich® Schlenk-type adapter
four-way adapter for solid-transfer vessel, male Joints: ST/NS 14/20 (3), female joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
Joint
female joint: ST/NS14/20
male Joints: ST/NS14/20 (3)
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)