Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z202819

Sigma-Aldrich

Glass spatulas

Square-cut and angled-cut ends, diam. × L 8 mm × 200 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$121.00

$121.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$121.00

About This Item

UNSPSC Code:
41122403
NACRES:
NB.35

$121.00


Please contact Customer Service for Availability

diam. × L

8 mm × 200 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service