Key Documents
Z197114
Sigma-Aldrich
3M disposable respirator
organic vapor/acid gas, size S
About This Item
UNSPSC Code:
46182002
NACRES:
NB.35
Product Z197114 is not currently sold in your country. Contact Technical Service
size
S
packaging
pkg of 1 ea
manufacturer/tradename
3M 5000
General description
Pre-assembled disposable respirators. Low-profile design provides improved comfort and less vision interference. Approved for: Organic vapor/acid gas and dusts/mists having a TWA not less than 0.05 mg/m3 or 2 mppcf.
NIOSH approved.
Protection factor: Up to 10 × PEL.
Legal Information
3M is a trademark of 3M Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)