Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z183806

SoftGUARD® hand cream

conditions the skin to prevent and relieve chapping and irritation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

General description

Provides barrier protection from harsh solutions. Conditions the skin to prevent and relieve chapping and irritation.

Packaging

3 oz tube

Legal Information

SoftGUARD is a registered trademark of Stahmer, Weston, & Co., Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service