Key Documents
Z180165
Supelco
End-fitting adapter for large columns
NPTF 1/2 in.
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
manufacturer/tradename
Ace Glass 585720
technique(s)
LPLC: suitable
NPTF
1/2 in.
General description
Polypropylene. Connects tubing adapters with NPT threads to column. Supplied with a locking retainer to hold 350 μm support screen (included) firmly in place. All adapters have an AceThred #50 and include one FETFE® O-ring
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)