Key Documents
Z178365
Scienceware® dispensing jug
snap closure, prevents "glugging", pkg of 1 ea
Synonym(s):
5 gallon bottle, water containers, water storage
Select a Size
1 EA
$79.30
$79.30
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$79.30
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
$79.30
Please contact Customer Service for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art
container capacity
5 gal (20 L)
General description
Five-gallon HDPE jug has flat sides, rounded corners, carrying handle, and bottom grip for pouring. Fill opening is 52 mm and contains a screw-cap having center thread for spigot. LDPE spigot is 1/4 turn type with 5/8 in. diameter outlet tube (1 1/4 in. long).
Jug lies on side with spigot overhanging shelf or table. Air vent, with snap closure, prevents "glugging". Spigot extends 70 mm from cap. 1 gal-molded-in graduations. 25 cm2 × 38 cm high over the handle.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1