Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z170488

Sigma-Aldrich

Lightnin® heavy-duty ring stand

rod diam. × L 5/8 in. × 27 in. , 316 SS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.35

manufacturer/tradename

Lightnin 829453

rod diam. × L

5/8 in. × 27 in. , 316 SS

weight

14 lb

General description

Heavy base. Suitable for mounting stirring motors, etc.

Legal Information

Lightnin is a registered trademark of Lightnin, a unit of General Signal

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service