Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z170372

Lightnin® 316 stainless steel impeller

4 blade turbine, diam. 2.0 in. , overall

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.72

manufacturer/tradename

Model A-200
Lightnin 829448

bore size

5/16 in.

diam.

2.0 in. , overall

Looking for similar products? Visit Product Comparison Guide

General description

Has 45° pitch for semi-axial flow.

Legal Information

Lightnin is a registered trademark of Lightnin, a unit of General Signal

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service