Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z162515

Sigma-Aldrich

Wheaton PP autosampler vials, Snap Ring

volume 0.1 mL, natural polypropylene vial, 5 × 11 serum finish, thread for 11 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

material

natural polypropylene vial

feature

closure type snap ring vial

packaging

pkg of 100 ea

O.D. × H

12 mm × 32 mm

volume

0.1 mL

fitting

thread for 11 mm

General description

0.1 mL vol., 12 × 32 mm. Lightweight, nonbreakable, high chemical resistance. Compatible with Beckman®, Agilent®, PerkinElmer®, Spectra-Physics® autosamplers.
1PAK = 1,000 vials
0.1PAK = 100 vials

Legal Information

Agilent is a registered trademark of Agilent Technologies, Inc.
Beckman is a registered trademark of Beckman Coulter, Inc.
PerkinElmer is a registered trademark of PerkinElmer Inc.
Spectra-Physics is a registered trademark of Newport Corporation

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service