Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z134120

Whatman® weighing boats and paper

B-2 Sheets, 4 x 4 inch, 500/pk

Synonym(s):

weigh paper, weighing paper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812

material

paper
square

feature

grade B-2

packaging

pack of 500

manufacturer/tradename

Whatman 10347672
Whatman Article No. 28414663 (US reference)

size

4 in. × 4 in.

Looking for similar products? Visit Product Comparison Guide

Other Notes

Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.

The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.

Legal Information

Whatman is a registered trademark of Cytiva

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service