Key Documents
Z127868
Sigma-Aldrich
Aldrich® large separatory funnel
overall capacity 6 L, PTFE stopcock, joint: ST/NS 45/40
Select a Size
1 EA
$1,740.00
$1,740.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
$1,740.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
$1,740.00
Please contact Customer Service for Availability
material
PTFE stopcock
Joint
joint: ST/NS45/40
capacity
6 L
Related Categories
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)