Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

Z127361

Cochranes Minit size bond tube sets

assortment set

Synonym(s):

chemistry model sets, molecular model sets

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
60104506
NACRES:
NB.17
Pricing and availability is not currently available.

manufacturer/tradename

Cochranes 0106

Looking for similar products? Visit Product Comparison Guide

General description

Components:
green, 2.5 cm (160)
green, 2 cm (160)
green, 1.5 cm (400)
green, 21 cm, (10)
white, 21 cm (6)
red, 21 cm (3)
flexible white, 21 cm (5)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service