Key Documents
Z126454
Automatic shut-off quick-disconnect
coupling bodies, tube to pipe thread, bore 1/4 in.
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
manufacturer/tradename
CRC Press
bore
1/4 in.
General description
Spring-loaded, locking, made of PP with 316 SS springs and EPR seals. When pulled apart, both sides seal quickly and automatically, making couplings excellent for handling air-sensitive liquids. Available for flexible tubing or rigid PE and nylon tubing using a ferruleless nut. Max. pressure: 90 psi at 70°F. Working temp.: 32 to 230°F (air/water). Use same bore size for coupling insert and body.
Choose from one of the most recent versions:
Certificates of Analysis (COA)