Key Documents
Z117447
LDPE laboratory tubing
I.D. × O.D. 3/16 in. × 5/16 in.
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
I.D. × O.D.
3/16 in. × 5/16 in.
wall thickness
1/16 in.
General description
Good chemical resistance, translucent, and rigid. Temp. range: -103 to 120°F.
Choose from one of the most recent versions:
Certificates of Analysis (COA)