Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z113999

Sigma-Aldrich

Wheaton glass serum bottle

volume 50 mL, clear, O.D. 20 mm, diam. × H 43 mm × 73 mm

Synonym(s):

Glass Serum Vials

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
$83.90
288 EA
$1,050.00

$83.90


Available to ship onApril 23, 2025Details


Request a Bulk Order

Select a Size

Change View
12 EA
$83.90
288 EA
$1,050.00

About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

$83.90


Available to ship onApril 23, 2025Details


Request a Bulk Order

material

borosilicate glass bottle

description

USP Type I

feature

autoclavable
closure type crimp top vial

O.D.

20 mm

capacity

50 mL

diam. × H

43 mm × 73 mm

volume

50 mL

color

clear

fitting

opening for 20 mm

Looking for similar products? Visit Product Comparison Guide

General description

It is autoclavable, easy sterilization, is used to contain substances to prevent contamination.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service