Key Documents
Z111392
Sigma-Aldrich
Heavy-duty Wig-L-Bug® grinder/mixer analog
AC/DC input 115 V AC
About This Item
UNSPSC Code:
23151817
availability
not available in EU
General description
Useful for mixing and grinding that require constant and long periods of operation. Has a 30 minute timer and fan-cooled motor. Comes with vial adapter for 1/2 in. diam. × 1 in. long vials.
Legal Information
Wig-L-Bug is a registered trademark of Dentsply Rinn
Choose from one of the most recent versions:
Certificates of Analysis (COA)