Key Documents
Z108294
Sigma-Aldrich
Tempo-Shield heat-resistant open top shield
pack of 1 pair
Select a Size
1 PKG
$87.10
$87.10
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
$87.10
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
$87.10
Please contact Customer Service for Availability
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Sigma-Aldrich
General description
Optional, open-top shield (mitten) allows for additional short-term, high-temperature (up to 1,000 °F), exposure protection while permitting finger dexterity.
Choose from one of the most recent versions:
Certificates of Analysis (COA)