Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z105902

Sigma-Aldrich

Cyanoacrylate adhesives

High-viscosity

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
31201600
NACRES:
NB.35

description

Ethyl Cyanoacrylate

viscosity

1300-1700 cP

Looking for similar products? Visit Product Comparison Guide

General description

  • Fast setting
  • Chemically resistant
  • Supplied in 1 oz bottles
  • High strength
  • Bond similar and dissimilar materials
  • Temp. range: -80 to 180 °C
Greatest gap filling; allows repositioning after assembly.

Physical properties

Shear Strength: 1800psi

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natta Wiriyakun et al.
Scientific reports, 6, 39115-39115 (2016-12-14)
Limitations of achieving highly sensitive and stable surface-enhanced Raman scattering (SERS) substrate greatly concern the suitable method for fabrication of large-area plasmonic nanostructures. Herein we report a simple approach using template-based synthesis to create a highly ordered two-dimensional array of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service