Documents
Z101249
Sigma-Aldrich
Aldrich® round-bottom flask with septum-inlet
capacity 250 mL, joint: ST/NS 19/22
Synonym(s):
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
single-neck
Joint
joint: ST/NS19/22
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
250 mL
General description
With ground-glass joint. Use septum Z565695 or Z565679 (see Septa section).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC