Key Documents
Y906506
Sigma-Aldrich
Valve
stainless steel bellows, 1/4 in. NPT straight path, external
About This Item
UNSPSC Code:
12142200
NACRES:
NA.12
Note: This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
material
stainless steel bellows
Quality Level
parameter
1000 psi max. pressure
L
1.9375 in.
weight
133 g
fitting
1/4 in. NPT straight path, external
Other Notes
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Choose from one of the most recent versions:
Certificates of Analysis (COA)