Skip to Content
MilliporeSigma
All Photos(1)

Documents

WHA77073200

Whatman® 850-DS 8 channel filter plates

0.45 μm, polyethersulfone membrane, non-sterile, 50 ea

Synonym(s):

disposable filter plate, dissolution sampling

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NC.01

product name

Whatman® 850-DS 8-channel filter plate, polyethersulfone membrane, pore size 0.45 μm, pack of 50 ea

material

polyethersulfone
polyethersulfone membrane

sterility

non-sterile

packaging

pack of 50 ea

manufacturer/tradename

Whatman 7707-3200
Whatman Article No. 29-0849-53 (US reference)

pore size

0.45 μm

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Save time and eliminate errors associated with manual sampling. Use Whatman 850-DS 8-channel filter plates in your Agilent 850-DS Dissolution Sampling Station

Application

The filter plates are exclusively designed for use with the optional filter module on the Agilent 850-DS Dissolution Sampling Station to simplify filter replacement between timepoints. Reliable alignment of the liquid path increases productivity in two ways. One, by reducing the risk of jamming, and two, by reducing leaks that may occur with manual sampling or other dissolution sample preparation systems.

Features and Benefits

  • Automated processing: up to 8 samples simultaneously
  • Readily available: in a wide range of pore sizes and materials

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service