Skip to Content
MilliporeSigma
All Photos(4)

Documents

WHA77013250

Whatman® UNIPLATE microplates

96 well, "V" bottom, well volume 250 μL, white polystyrene

Synonym(s):

Z746703, micro plate, multiwell plate, well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.24

material

V-bottom wells
white polystyrene

sterility

non-sterile

packaging

pkg of 50 ea

manufacturer/tradename

Whatman 7701-3250
Whatman Article No. 28420863 (US reference)

technique(s)

PCR: suitable

size

96 well

well volume

250 μL

working volume

250 μL

Looking for similar products? Visit Product Comparison Guide

General description

Cytiva′s Whatman UNIPLATE features a pointed bottom for high efficiency recovery of valuable, small volume samples. The vertical sides of the well ensure that material runs into the well base. Cytiva′s business offers several Whatman UNIPLATE options for sample collection and analysis. 24, 48, 96, and 384 well microplate options are available, as well as rectangular or round well bottom choices. Liquid recovery is necessary for a variety of applications that require samples to be re-analyzed. Collection of liquid samples is a necessary step in returning samples to a process line. Whatman UNIPLATE polystyrene microplates feature a ‘V’ well bottom well suited for recovering small volume liquid samples.

Application

  • Sample storage
  • Assay development
  • High-throughput screening
  • Plasmid miniprep
  • ELISA assays
  • Luminescence/chemiluminescence
  • Cell culture
  • Filtrate collection

Features and Benefits

  • ‘V′ bottom ensures maximum sample recovery – 99% liquid sample recovery is typical
  • Appropriate for applications with small, expensive sample volumes
  • High-quality polystyrene microplates
  • Suitable for use with robotic handlers and centrifuge carriers

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service