Skip to Content
MilliporeSigma
All Photos(1)

Documents

WHA77009904

Whatman® UNIFILTER® 24 well filter plates

25-30 μm, natural polypropylene, melt brown polypropylene membrane, non-sterile, biomolecule, 25 ea

Synonym(s):

Z746592, micro plate, multiwell plate, well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104900
NACRES:
NB.24

product name

Whatman® UNIFILTER® plates 24, 25 to 30 μm melt blown polypropylene, natural polypropylene, filter bottom with long drip director

material

polypropylene
polypropylene membrane

feature

25 to 30 μm melt blown polypropylene, natural polypropylene, filter bottom with long drip director

packaging

pkg of 25 ea

manufacturer/tradename

Whatman 7700-9904
Whatman Article No. 28420848 (US reference)

size

24 well

well maximum volume

1 μm

working volume

10 mL

pore size

25-30 μm

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

The 10 mL UNIFILTER is used with large sample or reagent volumes. Typical applications include biomolecule purification by solid phase extraction and organic synthesis in combinatorial chemistry library generation.

Polypropylene microplate for biomolecule purification

Applications such as combinatorial chemistry library generation that require large sample volumes depend on microplates capable of filtering and holding these volumes. Biomolecule purification by solid phase extraction (SPE) is an example of one such an application.

Whatman UNIFILTER 24 well microplates are well suited for large sample volume filtration. As SPE plates, they possess high heat and chemical resistance, as well as novel long drip directors that minimize the risk of well-to-well cross-contamination.

Features and Benefits

  • Polypropylene microplate is heat and chemical resistant
  • 24 well microplate with 10 mL wells appropriate for large sample volumes
  • No cross talk: patented integral filter drip director design prevents well-to-well cross contamination
  • Filtration media options include glass microfiber, melt-blown polypropylene, PTFE, VFE

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service