Skip to Content
MilliporeSigma
All Photos(2)

Documents

WHA7592104

Whatman® PTFE PM2.5 filter discs 2 µm pore size

white, hydrophobic, PP support ring, 50 ea, 46.2 mm diam

Synonym(s):

PTFE Filter, Z746347, air sampling filters

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24

product name

Whatman® PM 2.5 air monitoring membrane,

sterility

non-sterile

packaging

pkg of 50 ea

manufacturer/tradename

Whatman 7592-104
Whatman Article No. 28420782 (US reference)

application(s)

air monitoring

General description

Cytiva′s Whatman PM2.5 PTFE membrane filtration devices are designed for PM2.5 ambient air testing during continuous monitoring. These high-purity, thin PTFE membrane filters are sequentially numbered and come with chemically resistant polypropylene support rings.

PTFE membrane filters offer high chemical resistance and mechanical strength

One of the major applications for the PTFE membranes is the clarification of corrosives, solvents, and aggressive fluids. This includes the important requirement in HPLC analysis for sample filtration where any solid particles can cause permanent damage to the column. Air and gas sterilization make use of the hydrophobic characteristics of PTFE membrane filters and their ability to stop aqueous aerosols.

Robotic air sampling for PM2.5 air monitoring

Continuous air monitoring methods require a chemically resistant air sampling filter material that can withstand environmental pollution potentially containing a variety of contaminants and can still deliver accurate results. PM2.5 PTFE membrane filters from Cytiva business also feature a thermally stable design, making them ideal for ambient air monitoring. These membranes come in a polypropylene support designed for use in robotic air sampling of ambient air pollution.

PM2.5 air monitoring filter conformance

These air sampling filters conform to the PTFE Filters for EPA PM2.5 Reference Method, under the requirements of 40 CFR Part 50, Appendix L.

Features and Benefits

  • Polypropylene (PTFE) support rings for robotic air sampling and monitoring
  • Thermally stable design eliminates curling, keeps the membrane material flat, and makes the filter robot-friendly
  • Sequentially numbered filtration devices
  • Low tare mass for accurate gravimetric analysis and determinations

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service